Learn More
Heidolph™ Stirring Bar Sets
Shop All Heidolph Instruments Products
Click to view available options
Dimensions (L x W x H):
15 x 6 mm
25, 40, 50 mm
30 x 10 mm
Shape:
Cylinder
Oval
Unit Size:
Each
Pack of 3
Description
Oval Stir Bars:
- Shape is similar to that of the flask and ensures complete mixing
- Equipped with an oval magnetic core, more force is available than with a cylindrical magnetic core
- For applications in:
- Round bottom flasks e.g. for reaction mixtures
Cylindrical Stir Bars:
- Smooth operating conditions at optimum contact area
- Long lifetime
- For applications in:
- Beakers
- Crystallizing dishes
- Heating bathes
- Other flat bottom vessels
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Dimensions (L x W x H) | 25, 40, 50 mm |
| Coating | PTFE |
| Length (Metric) | 25, 40, 50 mm |
| Material | PTFE |
| Quantity | 1/Pk. |
| Shape | Cylinder |