Learn More
Heidolph™ Stirrer Guide (NS 29/32 PTFE)
Suitable for vacuum, perfect bearing for stainless steel and glass stirrer shafts
Brand:  Heidolph™ 509-09000-00
Product Code. 10637812
  • € 136.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Description
  • Ground joint cone made of PTFE with sealing rings on the outside to prevent sticking of the connection and to reduce danger of breaking glass
  • A special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap provides good sealing of the stirrer shaft
  • Accepts o.d. 8 mm shafts
Specifications
Specifications
Suitable for vacuum, perfect bearing for stainless steel and glass stirrer shafts
Stirrer Guide
PTFE