Learn More
Sampling Systems™ SteriWare™ Disposable LiquiThief
BIOPHARMA
Product Code. 15714758 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
290 mL
Unit Size:
Pack of 20
Product Code. 15714758
Brand: Sampling Systems™ 8050H1500
Sampling Systems™ SteriWare™ Disposable LiquiThief ideal for use with low viscocity liquids.
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • At the required depth pull up the handle
  • The further the handle is pulled, the larger the sample volume
  • Push handle down to expel the sample
  • No cross contamination
  • No cleaning required
Specifications
Capacity (Metric) 290 mL
Diameter (Metric) Outside 21 mm
Description Non-sterile, Individually Bagged
Length (Metric) 1500 mm
No. per Case 20