Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
BIOPHARMA
Product Code. p-7138878 Shop All Sampling Systems Products
Click to view available options
Diameter (Metric)Top:
100 mm
200 mm
250 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
Product Code. 16269421
Brand: Sampling Systems™ 8370H100S
Only null left
Add to basket
Add to basket
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
  • Constructed of HDPE
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Diameter (Metric)Top 100 mm
Diameter (Metric) Stem 12 mm
Sterility Sterile
Product Type Liquid Funnel