Learn More
Sampling Systems™ SteriWare™ Disposable Ladle
Shop All Sampling Systems Products
Click to view available options
Color:
Blue
White
Material:
HDPE
Polypropylene
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 40
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- The one piece moulded design has been specially constructed to go through a standard lid bung
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
Specifications
| Material | HDPE |
| Capacity (Metric) | 50 mL |
| Disposable | Single-use |
| Sterility | Sterile |
| Color | White |
| Diameter (Metric) Outer | 48 mm |