Learn More
Sampling Systems™ SteriWare™ Disposable Ladle
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Disposable Ladle allows for efficient surface sampling of liquids.
Brand: Sampling Systems™ 8053H-50
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- The one piece moulded design has been specially constructed to go through a standard lid bung
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
HDPE | |
Single-use | |
White |
50 mL | |
Non-sterile | |
48 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
121532 | Certificate of Compliance | 20/01/2025 | 8053H-50 |
121412 | Certificate of Compliance | 15/01/2025 | 8053H-50 |
120483 | Certificate of Compliance | 15/11/2024 | 8053H-50 |
119383 | Certificate of Compliance | 16/09/2024 | 8053H-50 |
117368 | Certificate of Compliance | 13/03/2024 | 8053H-50 |