Learn More
Thermo Scientific™ Sterilin™ Plain Flocked Swabs, Regular nylon flocked swab
Optimize your samples with our innovative nylon fiber technology flocked swabs. Thermo Scientific™ Sterilin™ Plain Flocked Swabs provide both outstanding collection and up to 95% release.
Brand: Thermo Scientific™ 502CS01
Description
Optimize your samples with our innovative nylon fiber technology flocked swabs. Thermo Scientific™ Sterilin™ Plain Flocked Swabs provide outstanding collection. Sample release from the flocked swab is at least 95% compared with 25% with a regular fiber swab, which is critical if the sample to be collected is particularly small.
- PCR compatible for molecular analysis
- Manufactured from completely inert components
- Certified RNase/DNase-free
- For forensic purposes product is available certified human DNA free and with a molded breakpoint so that it can be easily broken into a 2mL collection vial
- Available either in a paper peel pouch or in a plain dry tube
- CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
- Available in UK only
Specifications
| Regular nylon flocked swab | |
| Plastic | |
| Nylon | |
| Individually wrapped in paper peel pouch | |
| 80mm |
| Irradiated | |
| CE Marked as Class IIa in accordance with the Medical Device Directive93/42/EEC (for transient invasive use); Certified Human DNA-free, RNase/DNase-free | |
| Regular Flocked | |
| 100 | |
| Flocked swabs - Individually wrapped in paper peel pouch |