Learn More
Fisherbrand™ Sterile Syringes for Single Use
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
60 mL
Connection Type:
Leur Slip
Luer Lock
missing translation for 'unitSize'
Pack of 100
Pack of 50
Specifications
Specifications
| Diameter (Metric) Inner | 12.36 mm |
| Product Type | Luer-Lock |
| Volume (Metric) | 5 mL |
| Connection Type | Luer Lock |
| Sterility | Sterile |