Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
€ 490.00 - € 841.00
Specifications
Sterility | Sterile |
---|---|
Product Line | Wheaton CompletePAK |
Certifications/Compliance | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
Product Code | Brand | Closure Size | Style | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Style | Price | Quantity & Availability | ||||||
16227290
|
DWK Life Sciences
W986003-S |
13 mm | OmniFlex 3G Straight Plug | N/A | N/A | N/A | |||||
16257290
|
DWK Life Sciences
W986006-S |
20 mm | OmniFlex 3G Straight Plug | N/A | N/A | N/A | |||||
16237290
|
DWK Life Sciences
W986004-S |
13 mm | Ultra Pure Straight Plug | N/A | N/A | N/A | |||||
16267290
|
DWK Life Sciences
W986007-S |
20 mm | Ultra Pure Straight Plug | N/A | N/A | N/A | |||||
16247290
|
DWK Life Sciences
W986005-S |
13 mm | OmniFlex 3G Igloo | N/A | N/A | N/A | |||||
16277290
|
DWK Life Sciences
W986008-S |
20 mm | OmniFlex 3G Igloo | N/A | N/A | N/A | |||||
Description
- Sterile
- Stopper
Specifications
Sterile | |
Wheaton CompletePAK | |
USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers