Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
Click to view available options
Product Type:
Reducing Sleeve
Reflux Head
Small Volume Test Tube (3 mL)
Small Volume Test Tubes (1 mL)
Unit Size:
Each
Description
- Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations
- Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software
Specifications
Specifications
| Product Type | Reflux Head |
| For Use With (Application) | Reflux/Inert Head for 6 x 56 mm Dry Block |
| For Use With (Equipment) | For models 42754/55/56 |