Learn More
Bode Chemie™ Stellisept Med™ Antimicrobial Washing Lotion
Use as a skin friendly alternative to conventional decontamination preparations. Bode Chemie™ Stellisept Med™ Antimicrobial Washing Lotion is suitable for antimicrobial body cleansing and MRSA eradication in accordance with the Robert Koch-Institute recommendation.
Brand: Bode Chemie™ 9805012
Description
- Mild antimicrobial rinse-off wash lotion
- Comprehensive activity against bacteria within 30 seconds incl. MRSA/ ORSA and VRE
- Pleasant scent
- No alcohol and colorants
Specifications
Liquid | |
Clear | |
Bottle |
5.5 to 6.5 | |
Aqua, PEG–40 hydrogenated castor oil, PEG-150 distearate, glycerin, panthenol, PEG-7 glyceryl |