Learn More
Usbeck Laborgeraete™ Steel Triangles
Click to view available options
Length (Metric):
40 mm
50 mm
60 mm
80 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Each |
| Shape | Triangles |
| Length (Metric) | 60 mm |
| Material | Steel |
| Type | Wire Gauze |