accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Waste Disposal
Flammable Materials Waste Disposal Containers
Justrite™ Steel Safety Can
11973549
Print
Justrite™ Steel Safety Can
Capacity: 1L
Brand:
Justrite™ 10111Z
View more versions of this product
Product Code.
11973549
€ 199.00
/ Each
Quantity
Estimated Shipment: 07-01-2025
Log in
to see stock.
Explore more special offers
Certificates
Accessories and Services
Special Offers
Specifications
Capacity (Metric)
1 L
Diameter (Metric) Outer
122 mm
Height (Metric)
210 mm