Learn More
Fisherbrand™ Stainless Steel Straight Tongs
Product Code. 10409932 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
150 mm
200 mm
Unit Size:
Each
Product Code. 10409932
Only null left
Add to basket
Add to basket
Get maximum results using laboratory tongs in high temperature chemical reactions. Made of durable stainless steel, these Clifton™ Stainless Steel Straight Tongs ensure precise control of grasping, lifting, turning, and transferring heat-resistant and other materials in a laboratory environment.
  • Laboratory straight tongs
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Designed for corrosion resistance and durability ideal in laboratory routines
  • Laboratory metalware consumables
  • Finest in corrosion-resistant 304 stainless steel
  • Provides corrosion resistance that prevents product contamination
Specifications
Quantity 1 Ea.
Length (Metric) 150 mm
Material Steel
Type Crucible Tongs