Learn More
Justrite™ Steel Bench Can
Brand:  Justrite™ 10385
 View more versions of this product
Product Code. 11963629
  • € 135.00 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Specifications
Steel
4 L
114 mm
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Justrite™ Steel Bench Can > Capacity: 4L