Learn More
Brady™ Static Dissipative Thermal Transfer Printable Labels
Printable, static dissipative polyester labels for Thermal Transfer Printers.
Brand:  Brady™ 029908
 View more versions of this product
Product Code. 11543664
  • € 175.00 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Material is UL and CSA approved with its respective ribbon
  • Ideal for Circuit Board Labeling
  • For smooth surfaces
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Gloss
Thermal Transfer
Circuit Board Labeling, Electrical Labeling, General Identification
15.2 mm
10,000 Labels
-40°C to +120°C
9.5 mm
12.7 mm
7 mm
THT-96-473-10
White
Static-dissipative Polyester
Non-sterile
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
4
Rectangle
0.09 mm
12.7 mm
7 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers