Learn More
Abeba™ Static Control 31392 Shoes
ESD dissipative and Sanitized™ treated.
Brand: Abeba™ 31392/37
This item is not returnable.
View return policy
Description
- CE EN ISO 20345:2011 S2 SRA
- Microfiber
- White
- Composite toe cap
- PU midsole
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with elastic band
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Microfiber | |
Slip-resistant | |
31392 |
White | |
37 | |
Rubber | |
Composite |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Abeba™ Static Control 31392 Shoes > Size: 37