Learn More
Ismatec™ Standard Tubing, Tygon E-LFL
Product Code. 15723329
Click to view available options
Tubing Size:
I.D. 1.6 mm, O.D. 4.8 mm
I.D. 3.2 mm, O.D. 6.4 mm
I.D. 4.8 mm, O.D. 8.0 mm
Unit Size:
Each
Product Code. 15723329
Brand: Ismatec™ SCE0391
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Durable, chemically resistant tubing available in a broad range of sizes.
  • DEHP free.
  • Longest service life of any clear Tygon tubing material.
  • Excellent choice where transparency and good chemical resistance is needed.
  • Broad chemical resistance.
  • Tasteless.
  • Extremely low particulate spallation.
  • Meets USP Class VI and FDA criteria.
  • Non-Aging.
Specifications
Certifications/Compliance FDA 21 CFR 175.300; US Pharmacopoea Class VI
Autoclavable Autoclavable
Diameter (Metric) Inner 4.8 mm
Material Tygon E-LFL
Diameter (Metric) Outer 8.0 mm
Overall Length (Metric) 7.5 m
Min. Operating Temperature (Metric) −50°C
Packaging Quantity 1 Roll
Resistance Properties Acids: Fair, Alkaline solutions: Fair, Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent Sterile media: Limited
Tubing Size I.D. 4.8 mm, O.D. 8.0 mm
Sterilization Method Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky. Gas sterilization with ethylene oxide. Not recommended for sterilization with radiation.
Color Transparent
Max. Operating Temperature (Metric) 74°C