Learn More
Sampling Systems™ Standard Steel Sample Splitter Riffle Boxes
Shop All Sampling Systems Products
Click to view available options
Width (Metric) Chute:
7 mm
10 mm
15 mm
20 mm
25 mm
30 mm
40 mm
50 mm
65 mm
75 mm
80 mm
No. of Chutes:
8
10
12
Unit Size:
Each
Description
- Simply drop the sample into the top and the sample is split in two
- Split sample drops into the riffle boxes underneath
- It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
- Available in a wide range of sizes
Specifications
Specifications
| Product Type | Riffle Box |
| Material | Steel |
| Capacity (Metric) | 1.24 L |
| No. of Chutes | 12 |
| For Use With (Application) | Dividing and Reducing Bulk Samples |
| Dimensions (L x W x H) | 165 x 114 x 66 mm |
| Width (Metric) Chute | 7 mm |