Learn More
Thermo Scientific™ Standard Plate Count Agar (APHA) (Dehydrated)
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
€ 197.60 - € 1750.00
Specifications
Certifications/Compliance | APHA, AOAC, PHLS, ISO 11133:2014 |
---|---|
Form | Powder |
Product Type | Agar |
Test Type | Standard Plate Count for Microorganism |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
10679554
|
Thermo Scientific™
CM0463B |
500 g |
€ 197.60
500g |
In Stock
Log in to see stock. |
||||||
13205719
|
Thermo Scientific™
CM0463T |
5 kg |
€ 1750.00
5kg |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||||
Description
Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.
- Meets formulation of APHA, AOAC and PHLS
- Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014
Specifications
APHA, AOAC, PHLS, ISO 11133:2014 | |
Powder | |
Agar | |
Standard Plate Count for Microorganism |