Learn More
Koettermann™ Standard Hanging Cabinet, 1500x780x350mm
Koettermann™ Standard Hanging Cabinet, 1500x780x350mm provides suitable storage to accommodate a variety of laboratory needs.
Brand:  Koettermann™ 307-00179
754.41 EUR valid until 2024-11-01
Use promo code "21705" to get your promotional price.
 View more versions of this product
Product Code. 15753092
  • € 999.42 / Each
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Description
  • Wall-mounted cabinet
  • One shelf
  • Side panels
Specifications
Specifications
Wall-mounted Cabinet
Sliding Glass Doors
2
350 x 1500 x 780 mm
Yes
1