missing translation for 'onlineSavingsMsg'
Learn More
Standard Buffer Borate, pH 9.225, NIST, Chem-Lab Product Code.: 11999262

Standard Buffer Borate, pH 9.225, NIST, Chem-Lab

Product Code. 11999262
1 L
Click to view available options
Quantity:
1 L
10 L
This item is not returnable. View return policy

Product Code. 11999262

Brand: Chem Lab CL03.0202.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For laboratory use, pH measurements, traceable to SRM from NIST

pH = 9.464 at 0°C
pH = 9.395 at 5°C
pH = 9.332 at 10°C
pH = 9.276 at 15°C
pH = 9.225 at 20°C
pH = 9.180 at 25°C
pH = 9.139 at 30°C
pH = 9.068 at 40°C
pH = 9.011 at 50°C
pH = 8.962 at 60°C
pH = 8.921 at 70°C
pH = 8.885 at 80°C

Chemical Identifiers

CAS 1303-96-4
Molecular Formula Na2B4O7•10H2O

Specifications

Density 1.01g/mL
pH 9.225
Quantity 1 L
Packaging PE
Traceability to NIST NIST traceable
Formula Weight 381.37g/mol
Chemical Name or Material Sodium tetraborate decahydrate solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Standard Buffer Borate, pH 9.225, NIST, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.