missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schneider Gerd™ Stainless Steel Wire Mesh with Frame
Use this mesh for a variety of applications. Schneider Gerd™ Stainless Steel Wire Mesh with Frame features a sturdy 4301 stainless steel frame and woven mesh made of 4841 stainless steel.
Brand: Schneider Gerd™ 2573
This item is not returnable.
View return policy
Description
- Wire mesh with frame
- Fireproof up to 1100°C
- For general lab use
Specifications
155 x 155 mm | |
Stainless Steel | |
155 mm |
155 mm | |
Wire Mesh |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction