missing translation for 'onlineSavingsMsg'
Learn More

Wuertt Celluloid™ Stainless Steel Wire Basket

€ 116.63 - € 196.00

Specifications

Product Type Basket
Material Stainless Steel
For Use With (Application) Storage and Transport
Quantity 1 Each
View More Specs

Products 2
Product Code Brand Depth (Metric) Price Quantity & Availability  
Product Code Brand Depth (Metric) Price Quantity & Availability  
10372661
View Documents
Wuertt Celluloid™
5200113
100 mm
€ 196.00
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
10519051
View Documents
Wuertt Celluloid™
5201313
200 mm
€ 116.63
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Basket
Stainless Steel
Storage and Transport
1 Each
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.