Learn More
Thermo Scientific™ Stainless Steel Unions, Tees and Crosses
Shop All Thermo Scientific ProductsDescription
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
- Absolute zero or low dead volume formats
- Includes two stainless steel nuts and ferrules
Specifications
Specifications
| Type | Union |
| Swept Volume | 0.836 μL |
| Includes | Union with 2 stainless steel nuts and ferrules |
| Description | 0.050 in. through hole |
| Material | Stainless Steel |
| For Use With (Equipment) | Upchurch Scientific/Parker Fittings |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.