Learn More
asecos™ Stainless Steel Transport Canister
Shop All asecos Products
Click to view available options
Weight (Metric):
2 kg
2.2 kg
2.75 kg
3.3 kg
3.8 kg
Unit Size:
Each
Description
- incl. multilingual safety sticker for content labelling, with aluminium screw cap, ventilation valve, NBR seals and flexible plastic outlet pipe (Ø approx. 20 mm)
- UN-approval for packaging groups I, II and III
- Stainless steel
Specifications
Specifications
| Capacity (English) | 2.6 gal. |
| Capacity (Metric) | 10 L |
| Color | Gray |
| For Use With (Application) | Safe storage and transport |
| Material | Stainless Steel |
| Quantity | 1 |
| Width (English) | 6.69 in. |
| Width (Metric) | 170.00 mm |
| Height (English) | 11.02 in. |
| Height (Metric) | 280.00 mm |
| Show More |
Product Content Correction
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion