Learn More
asecos™ Stainless Steel Transport Canister
Shop All asecos Products
Click to view available options
Weight (Metric):
2 kg
2.2 kg
2.75 kg
3.3 kg
3.8 kg
Unit Size:
Each
Description
- incl. multilingual safety sticker for content labelling, with aluminium screw cap, ventilation valve, NBR seals and flexible plastic outlet pipe (Ø approx. 20 mm)
- UN-approval for packaging groups I, II and III
- Stainless steel
Specifications
Specifications
| Capacity (English) | 2.6 gal. |
| Capacity (Metric) | 10 L |
| Color | Gray |
| Material | Stainless Steel |
| Width (English) | 9.61 in. |
| Width (Metric) | 244.00 mm |
| Height (English) | 7.32 in. |
| Height (Metric) | 186.00 mm |
| Weight (English) | 4.85 lb. |
| Weight (Metric) | 2.2 kg |