Learn More
Grant Instruments™ Stainless Steel Test Tube Rack
Click to view available options
For Use With (Equipment):
0.5mm dia. Tubes
1.5mm dia. Tubes
10mm dia. Tubes
13mm dia. Tubes
16mm dia. Tubes
19mm dia. Tubes
25mm dia. Tubes
30mm dia. Tubes
Unit Size:
Each
Description
Fits a variety of test tube sizes from 10 to 30mm diameter
- For use with LSB Aqua Pro
- Available in 8 sizes to hold up to 48 SR test tubes
Specifications
Specifications
| Capacity (Metric) | 12 mL |
| For Use With (Equipment) | 25mm dia. Tubes |
| Material | Stainless Steel |