Learn More
RETSCH Stainless Steel Test Sieves Intermediate Rings
Brand:  RETSCH 60.935.000400
 View more versions of this product
Product Code. 10563922
  • € 201.00 / Each
Estimated Shipment: 30-07-2025
to see stock.
Explore more special offers
Specifications
Test Sieve Intermediate Ring
400 mm
Horizontal Sieve Shaker AS 400 control
Stainless Steel
65 mm
400 mm dia. x 65 mm H
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RETSCH Stainless Steel Test Sieves Intermediate Rings > Size (dia. x H): 400 x 65mm