Learn More
Socorex™ Stainless Steel Straight Drenchers
Click to view available options
Length (Metric):
25 mm
51 mm
102 mm
203 mm
Unit Size:
Pack of 2
Pack of 3
Description
- Stainless Steel
- Various lengths to accommodate feeding needs
- Autoclavable
- Reusable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Drenching Cannula |
| Material | Stainless Steel |
| Length (Metric) | 203 mm |