Learn More
Interscience Laboratories BagMixer™ Paddle Blender Bag Storage
The BagMixer™ Paddle Blender Bag Storage Rack is a specially designed rack that allows you to easily place samples in open bags.
Brand: Interscience Laboratories 221040
This item is not returnable.
View return policy
Description
- Stackable.
- Autoclavable.
- All stainless steel.
- BagRack™ allows clean and methodical storage of bags which can be left open to pipet easily or can be closed with a BagClip™.
Specifications
Storage Rack | |
Blood Bag Holders (12) | |
Rectangular | |
For 12 Bags 400 mL |
Autoclavable | |
Stainless Steel | |
Yes | |
1 Ea. |
Product Content Correction
Product Title
Interscience Laboratories BagMixer™ Paddle Blender Bag Storage > BagMixer™ Paddle Blender Bag Storage; Holds: 12 x 400 mL Bags