Learn More
Fisherbrand™ Stainless Steel Stirring Shafts
For use with overhead stirrer units. Fisherbrand™ Stainless Steel Shafts help avoid spillages and ensure even, thorough stirring.
Brand: Fisherbrand™ A00001308
Description
- Available with either anchor or 6-hole paddle blades
- Economically priced
- Optimum chemical resistance
Specifications
Overhead Stirrer | |
1 Each | |
0.99 kg |
Paddle blade, 6 holes | |
Stirring Shaft | |
Stainless Steel |