Learn More
Huenersdorf™ Stainless Steel Spare Sieve Insert for 105mm Funnels
A replacement sieve insert for use with 105mm funnels.
Brand:  Huenersdorf™ 958300
Product Code. 10693675
  • € 6.90 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Wire gauge 0.16mm
  • Mesh size 0.42mm
  • Made of stainless steel
Specifications
Specifications
Stainless Steel
105mm
Drum Funnel 953100
0.42mm
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Huenersdorf™ Stainless Steel Spare Sieve Insert for 105mm Funnels > 105mm; sieve insert