Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN260plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SN260plusB29726
Product Code. 17578565
  • € 190.00 / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SN260plus Sterilizer
Drip Tray
Rim: 15 mm