Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN260plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand: Memmert™ SN260plusB29726
This item is not returnable.
View return policy
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
SN260plus Sterilizer | |
Drip Tray | |
Rim: 15 mm | |
Stainless Steel |