Learn More
VitLab™ Stainless Steel Sieve
Made from stainless steel. Vitlab™ Stainless Steel Sieve is used with funnels no. 41794, 41894.
Brand:  VitLab™ 42099
5.26 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 10211691
  • € 8.30 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Description
  • 50mm insert diameter
  • For use with Vitlab™ funnels no. 41794, 41894
Specifications
Specifications
50mm
Vitlab™ funnels no. 41794, 41894
1 Each
Stainless Steel
0.45mm
Sieve Strainer