Learn More
Bochem™ Stainless Steel Rods
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
1500 mm
Length (Metric) Rod:
500 mm
600 mm
750 mm
1000 mm
1500 mm
Unit Size:
Each
Description
- Stainless Steel
- Sturdy
Specifications
Specifications
| Material | Stainless Steel (18/10) |
| Length (Metric) | 500 mm |
| Length (Metric) Rod | 500 mm |
| Diameter (Metric) Rod | 12 mm |
| Product Type | Stainless Steel Rod |
| Thread Size | Without Thread |
| Description |
|
| For Use With (Equipment) | For stand bases |
| Quantity | 1 Ea. |