Learn More
Cole-Parmer™ Essentials Stainless Steel Pail
Shop All Cole-Parmer Products
Click to view available options
Unit Size:
Each
Description
- Minimize dripping with flat pouring rim
Specifications
Specifications
| Product Type | Pail |
| Material | Stainless Steel |
| Capacity (English) | 6.1 gal. |
| Diameter (English) Outer | 11.9 in. |
| Height (English) | 14.9 in. |
| Height (English) With Closure | 14.9 in. |
| Quantity | 1 Ea. |