Learn More
Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate
Product Code. 10215701 Shop All Sartorius Products
Click to view available options
Mass:
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
Each
Product Code. 10215701
Brand: Sartorius™ YCW714800
This item requires a price update prior to being ordered.
Request a Price for this item here.
Highly polished steel calibration weights from 100g to 10kg
To OIML Class E2, F1 and F2 specification which includes DKD (Deutscher Kalibrierdienst accredited by the PTB) certificate (UKAS Equivalent).
TRUSTED_SUSTAINABILITY
Specifications
Case Material Plastic
Mass 10 kg