Learn More
Aalborg™ Stainless Steel N2 Mass Flowmeter
Brand:  Aalborg™ GFC17A-VAL6-C0/3NC-09-SS
Product Code. 11757889
  • € 1495.00 / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Mass Flowmeter
NIST calibration certificate
12 V to 200 mA max
Full scale
±5%
0 to 5 LPM
±1.5 %
5°C to 50°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers