Learn More
Sampling Systems™ Stainless Steel Ladles
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
50 mL
120 mL
150 mL
Unit Size:
Each
Description
- Satin finish
- Crevice-free construction so they can be easily cleaned between batches
- Range of sizes available
Specifications
Specifications
| Product Type | Ladle |
| Material | Stainless Steel (AISI 304) |
| Capacity (Metric) | 120 mL |
| For Use With (Application) | Sampling liquid products |
| Length (Metric) | 345 mm |