Learn More
Bochem™ Stainless Steel Laboratory Pot Lid
Click to view available options
Diameter (Metric) Inner:
160 mm
180 mm
200 mm
220 mm
240mm
320 mm
Unit Size:
Each
Description
Includes
Lid only.
- Stainless Steel
- For use with Bochem pots 8360-67
Specifications
Specifications
| Diameter (Metric) Inner | 160 mm |
| Material | Stainless Steel (18/10) |
| Product Type | Laboratory Pot Lid |