Learn More
Schneider Gerd™ Stainless Steel Laboratory Forceps
Use these forceps for a variety of applications. Schneider Gerd™ Stainless Steel Laboratory Forceps feature a grooved body for optimal grip.
Brand: Schneider Gerd™ 113
This item is not returnable.
View return policy
Description
- Stainless Steel Laboratory Forceps
- Ideal for dissection and general lab applications
Specifications
Blunt | |
145 mm | |
Each |
Stainless Steel | |
Forceps | |
Forceps, Laboratory |
Product Content Correction
Product Title
Schneider Gerd™ Stainless Steel Laboratory Forceps > Tip Style: Blunt point; Length: 145mm