Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Shop All Nickel-Electro Products
Click to view available options
Capacity (Metric):
200 mL
350 mL
550 mL
700 mL
1000 mL
1250 mL
2000 mL
2500 mL
5000 mL
missing translation for 'unitSize'
Each
Pack of 5
Description
- Durable stainless steel laboratory beaker
- 304 spec stainless steel beakers
- Will not shatter or chip
- Suitable for laboratory applications since
- All surfaces are non-reacting
- Offer zero absorption
Specifications
Specifications
| Diameter (Metric) Outer | 100 mm |
| Capacity (Metric) | 1250 mL |
| Material | Stainless Steel |
| Height (Metric) | 162 mm |
| Includes | Spout |
| Type | Beaker |