Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
Brand: Bandelin Electronic™ 358
This item is not returnable.
View return policy
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
- Available for a wide range of tank capacities
- Available in a range of mesh sizes
- Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Basket Insert | |
455 mm | |
Stainless Steel | |
245 x 50 x 275 mm |
Ultrasonic Bath, RK 1028/H / DT 1028 / H, DL 1028 H | |
245 mm | |
50 mm | |
K28 |