Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
Brand:  Bandelin Electronic™ 358
 View more versions of this product
Product Code. 10324491
  • € 356.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
  • Available for a wide range of tank capacities
  • Available in a range of mesh sizes
  • Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
Basket Insert
455 mm
Stainless Steel
245 x 50 x 275 mm
Ultrasonic Bath, RK 1028/H / DT 1028 / H, DL 1028 H
245 mm
50 mm
K28
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers