Learn More
Sampling Systems™ Stainless Steel Industrial Scoop
Product Code. 15859701 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
100 mL
200 mL
300 mL
500 mL
Length (Metric):
180 mm
205 mm
230 mm
270 mm
Width (Metric):
60 mm
75 mm
90 mm
110 mm
Unit Size:
Each
Product Code. 15859701
Brand: Sampling Systems™ A643300
Only null left
Add to basket
Add to basket
Constructed of high quality stainless steel to make it low cost and easy to use. Sampling Systems™ Stainless Steel Industrial Scoop is ideal for general and pharmaceutical use.
  • Light duty construction
  • Stainless Steel
  • Fully welded construction
  • Easy to clean
Specifications
Capacity (Metric) 300 mL
Product Type Scoop
Finish Polished
Width (Metric) 90 mm
Material Stainless Steel
Length (Metric) 230 mm