Learn More
Usbeck Laborgeraete™ Stainless Steel Forceps
Click to view available options
Length (Metric):
105 mm
115 mm
130 mm
160 mm
200 mm
Design:
Curved
Straight
Unit Size:
Each
Specifications
Specifications
| No. per Pack | 1 Pack |
| Design | Curved |
| Material | Stainless Steel |
| Length (Metric) | 105 mm |
| Dimensions (L x W) | 105 L mm |
| Type | Forceps |