Learn More
RETSCH Stainless Steel Distance Sieve For Ultra Centrifugal Mill ZM 200
Brand:  RETSCH 03.647.0259
Product Code. 10142152
  • € 554.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Specifications
1Each
Distance Sieve
Ultra Centrifugal Mill ZM 200
1 mm
Stainless Steel
For Temperature-sensitive Materials