Learn More
Fisherbrand™ Stainless Steel Crucible Tongs
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
200 mm
305 mm
381 mm
508 mm
Unit Size:
Each
Specifications
Specifications
| Description | Bow pattern |
| Quantity | 1 Ea. |
| Tip Style | Bent |
| Length (Metric) | 381 mm |
| Material | Stainless Steel |
| Type | Crucible Tongs |