Learn More
Amsta Rostfritt Stainless Steel Containers With Lid and 3 Clamps
Product Code. p-8009089
Click to view available options
Volume (Metric):
1 L
2 L
3 L
5 L
6 L
8 L
10 L
20 L
Unit Size:
Each
Product Code. 11761934
Brand: Amsta Rostfritt 50603101+5051310X3
Only null left
Specifications
Product Type Container with Lid and 3 Clamps
Material Stainless Steel
Dimensions (L x W) 220 x 300 mm
Volume (Metric) 10 L